Invitrogen
Manufacturer Code:PA559601
Catalog # PIPA559601
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ATXN2L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A2D A2LG A2LP A2RP ataxin 2 related protein ataxin-2 domain protein ataxin-2-like protein ataxin-2-related protein; ataxin 2 related protein; ataxin 2-like; Ataxin-2 domain protein; Ataxin-2-like protein; Ataxin-2-related protein
Gene Aliases: A2D; A2LG; A2LP; A2RP; ATXN2L
UniProt ID: (Human) Q8WWM7
Entrez Gene ID: (Human) 11273
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.