Invitrogen
Manufacturer Code:PA5106608
Catalog # PIPA5106608
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ATXN2L corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A2D A2LG A2lp A2RP ataxin 2 like ataxin 2 related protein ataxin 2-like ataxin-2 domain protein Ataxin-2-like protein Ataxin-2-related protein Atxn2l Atxnl2; ataxin 2 related protein; ataxin 2-like; Ataxin-2 domain protein; Ataxin-2-like protein; Ataxin-2-related protein
Gene Aliases: A2D; A2LG; A2LP; A2RP; ATXN2L; Atxnl2
UniProt ID: (Human) Q8WWM7, (Mouse) Q7TQH0
Entrez Gene ID: (Human) 11273, (Mouse) 233871
Molecular Function: RNA binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.