Invitrogen
Manufacturer Code:MA527732
Catalog # PIMA527732
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT4212 |
Immunogen | Recombit protein encompassing a sequence within the C-terminus region of human ATRX. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alpha thalassemia/mental retardation syndrome (X-linked); alpha thalassemia/mental retardation syndrome X-linked (RAD54 homolog, S. cerevisiae); ATP-dependent helicase ATRX; DNA dependent ATPase and helicase; helicase 2, X-linked; Heterochromatin protein 2; HP1 alpha-interacting protein; HP1-BP38 protein; mental retardation, X-linked 52; Transcriptional regulator ATRX; X-linked helicase II; X-linked nuclear protein; XNP; Zinc finger helicase; Znf-HX
Gene Aliases: 4833408C14Rik; AI447451; ATR2; ATRX; DXHXS6677E; HP1-BP38; Hp1bp2; Hp1bp38; JMS; MRX52; MRXHF1; MRXS3; RAD54; RAD54L; SFM1; SHS; XH2; XNP; ZNF-HX
UniProt ID: (Human) P46100, (Mouse) Q61687
Entrez Gene ID: (Human) 546, (Mouse) 22589
Molecular Function:
DNA binding protein
DNA helicase
helicase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.