Novus Biologicals
Manufacturer Code:NBP250111AF594
Catalog # NP20111A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 39f |
Immunogen | Recombinant Protein |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: alpha thalassemia/mental retardation syndrome X-linked (RAD54 homolog, S. cerevisiae); alpha thalassemia/mental retardation syndrome X-linked alpha thalassemia/mental retardation syndrome X-linked (RAD54 (S. cerevisiae)homolog) alpha thalassemia/mental retardation syndrome X-linked (RAD54 homolog S.cerevisiae) ATP-dependent helicase ATRX ATR2 DNA dependent ATPase and helicase EC 3.6.1 EC 3.6.4.12 helicase 2 X-linked Juberg-Marsidi syndrome MGC2094 MRXHF1 RAD54 homolog RAD54L SFM1 SHS transcriptional regulator ATRX XH2RAD54 X-linked helicase II X-linked nuclear protein XNPZNF-HX Zinc finger helicase Znf-HX; ATP-dependent helicase ATRX; DNA dependent ATPase and helicase; helicase 2, X-linked; mental retardation, X-linked 52; Transcriptional regulator ATRX; X-linked helicase II; X-linked nuclear protein; XNP; Zinc finger helicase; Znf-HX
Gene Aliases: ATR2; ATRX; JMS; MRX52; MRXHF1; RAD54; RAD54L; SFM1; SHS; XH2; XNP; ZNF-HX
UniProt ID: (Human) P46100
Entrez Gene ID: (Human) 546
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.