Abnova
Manufacturer Code:H00000546K
Catalog # 89-205-364
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human ATRX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human ATRX peptide using ARM Technology.
For Research Use Only
Protein Aliases: alpha thalassemia/mental retardation syndrome X-linked (RAD54 homolog, S. cerevisiae); ATP-dependent helicase ATRX; ATR2 MGC2094 MRXHF1 RAD54 RAD54L SFM1 SHS XH2 XNP ZNF-HX; DNA dependent ATPase and helicase; helicase 2, X-linked; mental retardation, X-linked 52; Transcriptional regulator ATRX; X-linked helicase II; X-linked nuclear protein; XNP; Zinc finger helicase; Znf-HX
Gene Aliases: ATR2; ATRX; JMS; MRX52; MRXHF1; RAD54; RAD54L; SFM1; SHS; XH2; XNP; ZNF-HX
UniProt ID: (Human) P46100
Entrez Gene ID: (Human) 546
Molecular Function: DNA binding protein DNA helicase helicase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.