Invitrogen
Manufacturer Code:PA517265
Catalog # PIPA517265
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to central residues of human ATR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ataxia telangiectasia and Rad3 related; Ataxia telangiectasia and Rad3-related protein; ATR serine/threonine kinase; FRAP-related protein 1; FRAP-related protein 1 FRAP-related protein-1 MEC1 mitosis entry checkpoint 1 homolog Rad3 related protein ataxia telangiectasia and Rad3-related protein protein kinase ATR serine/threonine-protein kinase ATR; FRAP-related protein-1; MEC1, mitosis entry checkpoint 1, homolog; Serine/threonine-protein kinase ATR
Gene Aliases: ATR; FCTCS; FRP1; I79_000761; MEC1; SCKL; SCKL1
UniProt ID: (Human) Q13535
Entrez Gene ID: (Human) 545, (Dog) 100856315, (Chinese hamster) 100766256, (Mouse) 245000, (Rat) 685055
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
nucleic acid binding
nucleotide kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.