Invitrogen
Manufacturer Code:PA133014
Catalog # PIPA133014
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Human ATR peptide amino acids 2122-2380 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ataxia telangiectasia and Rad3-related protein; FRAP-related protein 1; FRAP-related protein-1; FRP1 MEC1 SCKL SCKL1; MEC1, mitosis entry checkpoint 1, homolog; Serine/threonine-protein kinase ATR
Gene Aliases: ATR; FCTCS; FRP1; I79_000761; MEC1; SCKL; SCKL1
UniProt ID: (Human) Q13535
Entrez Gene ID: (Human) 545, (Chinese hamster) 100766256
Molecular Function: kinase non-receptor serine/threonine protein kinase nucleic acid binding nucleotide kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.