R&D Systems
Manufacturer Code:AF4717SP
Catalog # AF4717SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human ATR Glu2405-Met2644 Accession # Q13535 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ataxia telangiectasia and Rad3 related Ataxia telangiectasia and Rad3-related protein EC 2.7.11.1 FRAP-related protein 1 FRP1 FRP1FRAP-related protein-1 MEC1 protein kinase ATR Rad3 related protein SCKL1 SCKLMEC1 mitosis entry checkpoint 1 homolog serine/threonine-protein kinase ATR; Ataxia telangiectasia and Rad3-related protein; FRAP-related protein 1; FRAP-related protein-1; MEC1, mitosis entry checkpoint 1, homolog; Serine/threonine-protein kinase ATR
Gene Aliases: ATR; FCTCS; FRP1; MEC1; SCKL; SCKL1
UniProt ID: (Human) Q13535
Entrez Gene ID: (Human) 545
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
nucleic acid binding
nucleotide kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.