Novus Biologicals
Manufacturer Code:NBP202249
Catalog # NBP202249
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4E5 |
Immunogen | Full length human recombinant protein of human ATP5B (NP_001677) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP synthase F1 subunit beta; ATP synthase subunit beta mitochondrial ATP synthase H+ transporting mitochondrial F1 complex beta polypeptide ATPMB ATPSBmitochondrial ATP synthase beta subunit EC 3.6.3 EC 3.6.3.14 MGC5231 mitochondrial ATP synthetase beta subunit; ATP synthase subunit beta, mitochondrial; epididymis secretory protein Li 271; mitochondrial ATP synthase beta subunit; mitochondrial ATP synthetase, beta subunit
Gene Aliases: ATP5B; ATP5F1B; ATPMB; ATPSB; HEL-S-271
UniProt ID: (Human) P06576
Entrez Gene ID: (Human) 506
Molecular Function: ATP synthase DNA binding protein anion channel cation transporter hydrolase ion channel ligand-gated ion channel nucleic acid binding receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.