Novus Biologicals
Manufacturer Code:NBP188905
Catalog # NBP188905
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:MTDNIPLQPVRQKKRMDSRPRAGCCEWLRCCGG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPase class II type 9A; ATPase class II type 9A ATPase type IV phospholipid-transporting (P-type) ATPase class II type 9A ATPIIAphospholipid-transporting ATPase IIA EC 3.6.3 EC 3.6.3.1 KIAA0611probable phospholipid-transporting ATPase IIA; ATPase type IV, phospholipid-transporting (P-type),(putative); ATPase, class II, type 9A; phospholipid-transporting ATPase IIA; Probable phospholipid-transporting ATPase IIA
Gene Aliases: ATP9A; ATPIIA; KIAA0611
UniProt ID: (Human) O75110
Entrez Gene ID: (Human) 10079
Molecular Function: cation transporter hydrolase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.