Novus Biologicals
Manufacturer Code:NBP15701920UL
Catalog # NBP15701920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ATP8B2 (ATPase class I type 8B member 2) The peptide sequence was selected from the N terminal of ATP8B2. Peptide sequence KTSKYNILTFLPVNLFEQFQEVANTYFLFLLILQLIPQISSLSWFTTIVP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 36/8-9 fusion protein with epitope for anti-lectin antibody; ATPase class I type 8B member 2; ATPase class I type 8B member 2 ATPIDprobable phospholipid-transporting ATPase ID DKFZp434M0219 EC 3.6.3 EC 3.6.3.1 KIAA1137ATPase class I type 8B member 2 phospholipid-transporting ATPase ID; ATPase, aminophospholipid transporter, class I, type 8B, member 2; ATPase, class I, type 8B, member 2; P4-ATPase flippase complex alpha subunit ATP8B2; Phospholipid-transporting ATPase ID; probable phospholipid-transporting ATPase ID
Gene Aliases: ATP8B2; ATPID; KIAA1137
UniProt ID: (Human) P98198
Entrez Gene ID: (Human) 57198
Molecular Function: cation transporter hydrolase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.