Invitrogen
Manufacturer Code:PA562707
Catalog # PIPA562707
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ATP8A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aminophospholipid translocase; ATPase class I type 8A member 1; ATPase II; ATPase, aminophospholipid transporter (APLT), class I, type 8A, member 1; Chromaffin granule ATPase II; P4-ATPase flippase complex alpha subunit ATP8A1; Phospholipid-transporting ATPase IA; probable phospholipid-transporting ATPase IA
Gene Aliases: ATP8A1; ATPASEII; ATPIA; ATPP2
UniProt ID: (Human) Q9Y2Q0
Entrez Gene ID: (Human) 10396
Molecular Function: cation transporter hydrolase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.