Proteintech
Manufacturer Code:215651AP150UL
Catalog # 50-173-3486
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ATP8A1 Fusion Protein Ag16071 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aminophospholipid translocase; ATP8A1 ATPase II ATPASEII ATPIA ATPP2 Chromaffin granule ATPase II; ATPase 8A1, aminophospholipid transporter (APLT), class I; ATPase 8A1, p type; ATPase class I type 8A member 1; ATPase II; ATPase, aminophospholipid transporter (APLT), class I, type 8A, member 1; Chromaffin granule ATPase II; P4-ATPase flippase complex alpha subunit ATP8A1; Phospholipid-transporting ATPase IA; probable phospholipid-transporting ATPase IA
Gene Aliases: AI481521; AI853962; APLT; Atp3a2; ATP8A1; ATPASEII; Atpc1; ATPIA; ATPP2; AW743152; AW822227; B230107D19Rik; ClassI
UniProt ID: (Human) Q9Y2Q0, (Mouse) P70704
Entrez Gene ID: (Human) 10396, (Rat) 289615, (Mouse) 11980
Molecular Function: cation transporter hydrolase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.