Novus Biologicals
Manufacturer Code:NB100361AF700
Catalog # NB100361Z
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal sequence near the C-terminus of human ATP7b. [UniProt# P35670] |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPase Cu(2+)- transporting beta polypeptide ATPase Cu++ transporting beta polypeptide Copper pump 2 copper-transporting ATPase 2 EC 3.6.3 EC 3.6.3.4 PWD WC1 WD Wilson disease-associated protein WNDATPase Cu++ transporting beta polypeptide (Wilson disease); ATPase, Cu(2+)- transporting, beta polypeptide; ATPase, Cu++ transporting, beta polypeptide; Copper pump 2; Copper-transporting ATPase 2; Wilson disease-associated protein; WND/140 kDa
Gene Aliases: ATP7B; PWD; WC1; WD; WND
UniProt ID: (Human) P35670
Entrez Gene ID: (Human) 540
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.