Invitrogen
Manufacturer Code:MA527719
Catalog # PIMA527719
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S62-29 |
Immunogen | Synthetic peptide amino acids 3-21 (cytoplasmic N-terminus) of human Copper-transporting ATPase2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP7B ATPase Cu++ transporting beta polypeptide ATPase Cu(2+) transporting beta polypeptide Copper pump 2 Copper transporting ATPase 2 PWD Toxic milk tx WC1 WD Wilson disease associated protein WND WND/140 kDa; ATPase, Cu(2+)- transporting, beta polypeptide; ATPase, Cu++ transporting, beta polypeptide; ATPase, Cu++ transporting, beta polypeptide (same as Wilson disease); Copper pump 2; Copper-transporting ATPase 2; PINA gene, promoter; pineal night-specific ATPase; toxic milk; Wilson disease-associated protein; Wilson disease-associated protein homolog; Wilson protein; WND/140 kDa
Gene Aliases: Atp7a; ATP7B; Hts; PINA; PWD; tx; WC1; WD; WND
UniProt ID: (Human) P35670, (Mouse) Q64446
Entrez Gene ID: (Human) 540, (Rat) 24218, (Mouse) 11979
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.