Invitrogen
Manufacturer Code:MA527720
Catalog # PIMA527720
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S60-4 |
Immunogen | Synthetic peptide amino acids 42-61 (cytoplasmic C-terminus) of human Copper- transporting ATPase1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP7A ATP 7A ATPase Cu transporting DSMAX FLJ17790 MC1 MC 1 MK MNK OHS Copper pump 1 Menke OTTHUMP00000062077 SMAX3 ATPase copper transporting alpha polypeptide ATPase Cu++ transporting alpha polypeptide (Menkes syndrome) Copper transporting ATPase 1 Cu++ transporting P type ATPase Menkes disease associated protein Menkes syndrome; ATPase, Cu++ transporting, alpha polypeptide; Copper pump 1; Copper-transporting ATPase 1; Cu++-transporting P-type ATPase; Menkes disease-associated protein; Menkes disease-associated protein homolog; Menkes protein
Gene Aliases: ATP7A; DSMAX; MC1; MK; MNK; SMAX3
UniProt ID: (Human) Q04656, (Mouse) Q64430, (Rat) P70705
Entrez Gene ID: (Human) 538, (Mouse) 11977, (Rat) 24941
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.