Novus Biologicals
Manufacturer Code:NBP259376C
Catalog # NBP259376C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S60-4 |
Immunogen | Synthetic peptide amino acids 42-61 (cytoplasmic C-terminus) of human Copper- transporting ATPase1 |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ATPase Cu++ transporting alpha polypeptide Copper pump 1 copper-transporting ATPase 1 Cu++-transporting P-type ATPase DSMAX EC 3.6.3 EC 3.6.3.4 MC1 Menkes disease-associated protein Menkes syndrome MK MNKFLJ17790 OHS SMAX3; ATPase, Cu++ transporting, alpha polypeptide; Copper pump 1; Copper-transporting ATPase 1; Cu++-transporting P-type ATPase; Menkes disease-associated protein
Gene Aliases: ATP7A; DSMAX; MC1; MK; MNK; SMAX3
UniProt ID: (Human) Q04656
Entrez Gene ID: (Human) 538
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.