Invitrogen
Manufacturer Code:PA5103110
Catalog # PIPA5103110
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human ATP7A corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP 7A ATP7A ATPase copper transporting alpha ATPase Cu++ transporting alpha polypeptide copper pump 1 Copper-transporting ATPase 1 Cu++-transporting P-type ATPase DSMAX FLJ17790 MC 1 MC1 Menke Menkes disease-associated protein menkes disease-associated protein homolog Menkes protein Menkes syndrome MK Mnk OHS OTTHUMP00000062077 SMAX3; ATPase, Cu++ transporting, alpha polypeptide; Copper pump 1; Copper-transporting ATPase 1; Cu++-transporting P-type ATPase; Menkes disease-associated protein; Menkes disease-associated protein homolog; Menkes protein
Gene Aliases: ATP7A; DSMAX; MC1; MK; MNK; SMAX3
UniProt ID: (Human) Q04656, (Mouse) Q64430, (Rat) P70705
Entrez Gene ID: (Human) 538, (Mouse) 11977, (Rat) 24941
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.