Proteintech
Manufacturer Code:161431AP150UL
Catalog # 50-173-0750
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ATP6V1G1 Fusion Protein Ag9217 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP6G ATP6G1 ATP6GL ATP6J ATP6V1G1 V ATPase 13 kDa subunit 1 V ATPase subunit G 1 Vma10; ATPase, H+ transporting, lysosomal (vacuolar proton pump); ATPase, H+ transporting, lysosomal (vacuolar proton pump), member J; ATPase, H+ transporting, lysosomal 13kD, V1 subunit G; ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G1; ATPase, H+ transporting, V1 subunit G; lysosomal 13kDa; V-ATPase 13 kDa subunit 1; V-ATPase subunit G 1; V-type proton ATPase subunit G 1; vacuolar ATP synthase subunit M16; vacuolar H(+)-ATPase subunit G 1; Vacuolar proton pump subunit G 1; Vacuolar proton pump subunit M16
Gene Aliases: 1810024D14Rik; AA960677; ATP6G; ATP6G1; ATP6GL; ATP6J; ATP6V1G1; VAG1; Vma10
UniProt ID: (Human) O75348, (Mouse) Q9CR51
Entrez Gene ID: (Human) 9550, (Mouse) 66290, (Rat) 298103
Molecular Function:
ATP synthase
cation transporter
hydrolase
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.