Novus Biologicals
Manufacturer Code:NBP270238
Catalog # NBP270238
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E11 |
Immunogen | Full length human recombinant protein of human ATP6V1B2 (NP_001684) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: 000 subunit 56/58kD isoform 2 ATP6B2ATPase H&plus transporting lysosomal (vacuolar proton pump) beta polypeptide ATPase H&plus transporting lysosomal 56/58kDa V1 subunit B2 Endomembrane proton pump 58 kDa subunit H&plus transporting two-sector ATPase HO57ATP6B1B2 vacuolar H&plus-ATPase 56 Vacuolar proton pump subunit B 2 VATB V-ATPase B2 subunit V-ATPase subunit B 2 Vma2 VPP3ATPase H&plus transporting lysosomal 56/58kDa V1 subunit B isoform 2 V-type proton ATPase subunit B brain isoform; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B2; Endomembrane proton pump 58 kDa subunit; H+ transporting two-sector ATPase; HO57; testicular secretory protein Li 65; V-ATPase B2 subunit; V-ATPase subunit B 2; V-type proton ATPase subunit B, brain isoform; vacuolar H+-ATPase 56,000 subunit; Vacuolar proton pump subunit B 2
Gene Aliases: ATP6B1B2; ATP6B2; ATP6V1B2; DOOD; HO57; VATB; Vma2; VPP3; ZLS2
UniProt ID: (Human) P21281
Entrez Gene ID: (Human) 526
Molecular Function: ATP synthase DNA binding protein anion channel cation transporter hydrolase ion channel ligand-gated ion channel nucleic acid binding receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.