Invitrogen
Manufacturer Code:PA552518
Catalog # PIPA552518
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human V-ATPase B2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATPase V1 subunit B2 ATP6B1B2 ATP6B2 HO57 VATB VPP3 Vma2; ATPase, H+ transporting, lysosomal (vacuolar proton pump), beta 56/58 kDa, isoform 2; ATPase, H+ transporting, lysosomal 56/58kD, V1 subunit B, isoform 2; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B2; ATPase, H+ transporting, V1 subunit B, isoform 2; Endomembrane proton pump 58 kDa subunit; H+ transporting two-sector ATPase; HO57; lysosomal 56/58kDa; testicular secretory protein Li 65; V-ATPase B2 subunit; V-ATPase subunit B 2; V-type proton ATPase subunit B, brain isoform; vacuolar H+-ATPase 56,000 subunit; vacuolar H+ATPase B2; Vacuolar proton pump subunit B 2
Gene Aliases: AI194269; AI790362; ATP6B1B2; ATP6B2; ATP6V1B2; DOOD; HO57; R74844; Vat2; VATB; Vma2; VPP3; ZLS2
UniProt ID: (Human) P21281, (Mouse) P62814, (Rat) P62815
Entrez Gene ID: (Human) 526, (Mouse) 11966, (Rat) 117596
Molecular Function:
ATP synthase
DNA binding protein
anion channel
cation transporter
hydrolase
ion channel
ligand-gated ion channel
nucleic acid binding
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.