Abnova
Manufacturer Code:H00009114M01J
Catalog # 89-335-086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2G12 |
Immunogen | ATP6V0D1 (NP_004682 238 a.a. &sim 308 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody.
Protein Aliases: 32 kDa accessory protein; ATP6D ATP6DV P39 VATX VMA6 VPATPD; ATPase, H+ transporting, lysosomal (vacuolar proton pump), member D; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d1; H(+)-transporting two-sector ATPase, subunit D; p39; V-ATPase 40 KDa accessory protein; V-ATPase AC39 subunit; V-ATPase subunit d 1; V-ATPase, subunit D; V-type proton ATPase subunit d 1; Vacuolar proton pump subunit d 1
Gene Aliases: ATP6D; ATP6DV; ATP6V0D1; P39; VATX; VMA6; VPATPD
UniProt ID: (Human) P61421
Entrez Gene ID: (Human) 9114
Molecular Function:
ATP synthase
cation transporter
hydrolase
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.