Invitrogen
Manufacturer Code:PA545852
Catalog # PIPA545852
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ATP6V0D1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 32 kDa accessory protein; ATPase V0 subunit d1 ATP6D ATP6DV FLJ43534 P39 VATX VMA6 VPATPD; ATPase, H+ transporting, lysosomal (vacuolar proton pump), 42 kDa; ATPase, H+ transporting, lysosomal (vacuolar proton pump), member D; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit D; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d1; ATPase, H+ transporting, lysosomal V0 subunit d1; ATPase, H+ transporting, lysosomal, V0 subunit d1; ATPase, H+ transporting, V0 subunit D; cto; H(+)-transporting two-sector ATPase, subunit D; lysosomal 38kDa; P39; Physophilin; V-ATPase 40 KDa accessory protein; V-ATPase AC39 subunit; V-ATPase subunit d 1; V-ATPase, subunit D; V-type proton ATPase subunit d 1; Vacuolar proton pump subunit d 1; vacuolar proton-ATPase, subunit D; V-ATPase, subunit D
Gene Aliases: Ac39; AI267038; ATP6D; ATP6DV; ATP6V0D1; cto; fb73h07; P39; VATX; VMA6; VPATPD; wu:fb73h07; zgc:63769
UniProt ID: (Human) P61421, (Bovine) P61420, (Mouse) P51863, (Zebrafish) Q6PGV1
Entrez Gene ID: (Dog) 479685, (Rabbit) 100350157, (Human) 9114, (Bovine) 282148, (Horse) 100053316, (Guinea pig) 100733139, (Rat) 291969, (Mouse) 11972, (Zebrafish) 322811
Molecular Function: ATP synthase cation transporter hydrolase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.