Invitrogen
Manufacturer Code:PA5116996
Catalog # PIPA5116996
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-351 of human ATP6V0D1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 32 kDa accessory protein; 32 kDa accessory protein Ac39 AI267038 Atp6d ATP6DV Atp6v0d1 ATPase H+ transporting V0 subunit d1 ATPase H+ transporting lysosomal (vacuolar proton pump) 42 kDa ATPase H+ transporting lysosomal (vacuolar proton pump) member D ATPase H+ transporting lysosomal 38kDa V0 subunit D ATPase H+ transporting lysosomal 38kDa V0 subunit d1 ATPase H+ transporting lysosomal V0 subunit D1 ATPase H+ transporting V0 subunit D FLJ43534 H(+)-transporting two-sector ATPase subunit D lysosomal 38kDa P39 Physophilin Vacuolar proton pump subunit d 1 V-ATPase 40 KDa accessory protein V-ATPase AC39 subunit V-ATPase subunit d 1 V-ATPase subunit D VATX Vma6 VPATPD V-type proton ATPa V-type proton ATPase subunit d 1; ATPase, H+ transporting, lysosomal (vacuolar proton pump), 42 kDa; ATPase, H+ transporting, lysosomal (vacuolar proton pump), member D; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit D; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d1; ATPase, H+ transporting, V0 subunit D; H(+)-transporting two-sector ATPase, subunit D; lysosomal 38kDa; P39; Physophilin; V-ATPase 40 KDa accessory protein; V-ATPase AC39 subunit; V-ATPase subunit d 1; V-ATPase, subunit D; V-type proton ATPase subunit d 1; Vacuolar proton pump subunit d 1
Gene Aliases: Ac39; AI267038; ATP6D; ATP6DV; ATP6V0D1; P39; VATX; VMA6; VPATPD
UniProt ID: (Human) P61421, (Mouse) P51863
Entrez Gene ID: (Human) 9114, (Rat) 291969, (Mouse) 11972
Molecular Function:
ATP synthase
cation transporter
hydrolase
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.