Proteintech
Manufacturer Code:182741AP150UL
Catalog # 50-173-2083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ATP6V0D1 Fusion Protein Ag13002 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 32 kDa accessory protein; 32 kDa accessory protein ATP6D ATP6DV ATP6V0D1 P39 V ATPase AC39 subunit V ATPase subunit d 1 VATX VMA6 VPATPD; ATPase, H+ transporting, lysosomal (vacuolar proton pump), 42 kDa; ATPase, H+ transporting, lysosomal (vacuolar proton pump), member D; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit D; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d1; ATPase, H+ transporting, V0 subunit D; H(+)-transporting two-sector ATPase, subunit D; lysosomal 38kDa; P39; Physophilin; V-ATPase 40 KDa accessory protein; V-ATPase AC39 subunit; V-ATPase subunit d 1; V-ATPase, subunit D; V-type proton ATPase subunit d 1; Vacuolar proton pump subunit d 1
Gene Aliases: Ac39; AI267038; ATP6D; ATP6DV; ATP6V0D1; P39; VATX; VMA6; VPATPD
UniProt ID: (Human) P61421, (Mouse) P51863
Entrez Gene ID: (Human) 9114, (Rat) 291969, (Mouse) 11972
Molecular Function:
ATP synthase
cation transporter
hydrolase
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.