Novus Biologicals
Manufacturer Code:NBP159654
Catalog # NBP159654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human ATP6V0C (NP_001685). Peptide sequence VVAVLIANSLNDDISLYKSFLQLGAGLSVGLSGLAAGFAIGIVGDAGVRG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP6CATPase H+ transporting lysosomal (vacuolar proton pump) 16kD ATP6LVPPC ATPase H+ transporting lysosomal 16kDa V0 subunit c ATPL H(+)-transporting two-sector ATPase 16 kDa subunit vacuolar ATP synthase 16 kDa proteolipid subunit vacuolar H+ ATPase proton channel subunit Vacuolar proton pump 16 kDa proteolipid subunit VATL V-ATPase 16 kDa proteolipid subunit Vma3 V-type proton ATPase 16 kDa proteolipid subunit; ATPase, H+ transporting, lysosomal 16kDa, V0 subunit c; H(+)-transporting two-sector ATPase, 16 kDa subunit; V-ATPase 16 kDa proteolipid subunit; V-type proton ATPase 16 kDa proteolipid subunit; vacuolar ATP synthase 16 kDa proteolipid subunit; vacuolar H+ ATPase proton channel subunit; Vacuolar proton pump 16 kDa proteolipid subunit
Gene Aliases: ATP6C; ATP6L; ATP6V0C; ATPL; VATL; Vma3; VPPC
UniProt ID: (Human) P27449
Entrez Gene ID: (Human) 527
Molecular Function: ATP synthase cation transporter hydrolase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.