Invitrogen
Manufacturer Code:PA542156
Catalog # PIPA542156
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human ATP6V0A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A2V-ATPase; ATPase H+ transporting lysosomal V0 subunit a2 ATP6N1D ATP6a2 J6B7 Stv1 TJ6 TJ6M TJ6s Vph1 a2 A2 RTF WSS ARCL STV1 TJ6S VPH1 ARCL2A ATP6A2; ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 2 (38kD); ATPase, H+ transporting, lysosomal V0 subunit a; ATPase, H+ transporting, lysosomal V0 subunit a2; Immune suppressor factor J6B7; ISF; Lysosomal H(+)-transporting ATPase V0 subunit a2; regeneration and tolerance factor; ShIF; T-cell expressing clone j6; TJ6; V-ATPase 116 kDa; V-ATPase 116 kDa subunit a2; V-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a isoform 2; V-type proton ATPase 116 kDa subunit a2; vacuolar proton translocating ATPase 116 kDa subunit a; Vacuolar proton translocating ATPase 116 kDa subunit a isoform 2; vacuolar proton translocating ATPase 116-kDa subunit a2
Gene Aliases: 8430408C20Rik; A2; AI385560; ARCL; ARCL2A; ATP6A2; ATP6N1B; ATP6N1D; Atp6n2; ATP6V0A2; AW489264; C76904; Cc1-3; ISF; J6B7; RTF; SHIF; STV1; TJ6; TJ6M; TJ6S; V-ATPase 116 kDa; V-ATPase a2; VPH1; WSS
UniProt ID: (Human) Q9Y487, (Bovine) O97681, (Mouse) P15920
Entrez Gene ID: (Dog) 486245, (Horse) 100061140, (Pig) 100154525, (Human) 23545, (Bovine) 338038, (Mouse) 21871, (Guinea pig) 101787762, (Rat) 116455
Molecular Function:
ATP synthase
cation transporter
hydrolase
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.