Invitrogen
Manufacturer Code:PA5113233
Catalog # PIPA5113233
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human V-type proton ATPase 116 kDa subunit a isoform 1 protein (80-260AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a1 AA959968 adenosine triphosphatase ATP6a1 Atp6n1 ATP6N1A Atp6v0a1 ATPase H+ transporting lysosomal (vacuolar proton pump) noncatalytic accessory protein 1 (110/160 kDa) ATPase H+ transporting V0 subunit a1 ATPase H+ transporting lysosomal (vacuolar proton pump) noncatalytic accessory protein 1 (110/160 kDa) ATPase H+ transporting lysosomal (vacuolar proton pump) non-catalytic accessory protein 1A (110/116kD) ATPase H+ transporting lysosomal (vacuolar proton pump) noncatalytic accessory protein 1A (110/160 kDa) ATPase H+ transporting lysosomal non-catalytic accessory protein 1 (110/116kD) ATPase H+ transporting lysosomal noncatalytic accessory protein 1a ATPase H+ transporting lysosomal V0 subunit a ATPase H+ transporting lysosomal V0 subunit a1 Atpv0a1 BOS_19073 Clathrin-coated vesicle/synaptic vesicle proton pump 116 kDa subunit H(+)-transporting two-sector ATPase 116 kDa accessory protein A1 LOC100125982 Stv1 V-A Vacuolar adenosine triphosphatase subunit Ac116 Vacuolar proton pump subunit 1 vacuolar proton translocating ATPase 116 kDa subunit A Vacuolar proton translocating ATPase 116 kDa subunit a isoform 1 vacuolar proton-translocating ATPase a1 isoform vacuolar-type H(+)-ATPase 115 kDa subunit V-ATPase 116 kDa V-ATPase 116 kDa isoform a1 V-ATPase a1 v-H+ATPase subunit a1 Vph1 Vpp1 Vpp-1 V-type proton ATPase 116 kDa subunit a V-type proton ATPase 116 kDa subunit a isoform 1; ATPase H+ transporting lysosomal (vacuolar proton pump) noncatalytic accessory protein 1 (110/160 kDa); ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 1A (110/116kD); ATPase, H+ transporting, lysosomal (vacuolar proton pump) noncatalytic accessory protein 1 (110/160 kDa); ATPase, H+ transporting, lysosomal (vacuolar proton pump) noncatalytic accessory protein 1A (110/160 kDa); ATPase, H+ transporting, lysosomal non-catalytic accessory protein 1 (110/116kD); ATPase, H+ transporting, lysosomal noncatalytic accessory protein 1a; ATPase, H+ transporting, lysosomal V0 subunit a; ATPase, H+ transporting, lysosomal V0 subunit a1; Clathrin-coated vesicle/synaptic vesicle proton pump 116 kDa subunit; H(+)-transporting two-sector ATPase, 116 kDa accessory protein A1; V-ATPase 116 kDa; V-ATPase 116 kDa subunit a1; V-ATPase a1; v-H+ATPase subunit a1; V-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a isoform 1; V-type proton ATPase 116 kDa subunit a1; Vacuolar adenosine triphosphatase subunit Ac116; Vacuolar proton pump subunit 1; vacuolar proton translocating ATPase 116 kDa subunit A; Vacuolar proton translocating ATPase 116 kDa subunit a isoform 1; vacuolar-type H(+)-ATPase 115 kDa subunit
Gene Aliases: a1; AA959968; ATP6a1; ATP6N1; ATP6N1A; ATP6V0A1; Atpv0a1; Stv1; Vph1; Vpp-1; VPP1
UniProt ID: (Human) Q93050, (Mouse) Q9Z1G4, (Rat) P25286
Entrez Gene ID: (Human) 535, (Mouse) 11975, (Rat) 29757
Molecular Function:
ATP synthase
cation transporter
hydrolase
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.