Proteintech
Manufacturer Code:138281AP150UL
Catalog # 50-172-9028
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ATP6V0A1 Fusion Protein Ag4191 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a1 ATP6N1 ATP6N1A ATP6V0A1 Stv1 V ATPase 116 kDa isoform a1 Vacuolar proton pump subunit 1 Vph1 VPP1; ATPase H+ transporting lysosomal (vacuolar proton pump) noncatalytic accessory protein 1 (110/160 kDa); ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 1A (110/116kD); ATPase, H+ transporting, lysosomal (vacuolar proton pump) noncatalytic accessory protein 1 (110/160 kDa); ATPase, H+ transporting, lysosomal (vacuolar proton pump) noncatalytic accessory protein 1A (110/160 kDa); ATPase, H+ transporting, lysosomal non-catalytic accessory protein 1 (110/116kD); ATPase, H+ transporting, lysosomal noncatalytic accessory protein 1a; ATPase, H+ transporting, lysosomal V0 subunit a; ATPase, H+ transporting, lysosomal V0 subunit a1; Clathrin-coated vesicle/synaptic vesicle proton pump 116 kDa subunit; H(+)-transporting two-sector ATPase, 116 kDa accessory protein A1; V-ATPase 116 kDa; V-ATPase 116 kDa subunit a1; V-ATPase a1; v-H+ATPase subunit a1; V-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a isoform 1; V-type proton ATPase 116 kDa subunit a1; Vacuolar adenosine triphosphatase subunit Ac116; Vacuolar proton pump subunit 1; vacuolar proton translocating ATPase 116 kDa subunit A; Vacuolar proton translocating ATPase 116 kDa subunit a isoform 1; vacuolar-type H(+)-ATPase 115 kDa subunit
Gene Aliases: a1; AA959968; ATP6a1; ATP6N1; ATP6N1A; ATP6V0A1; Atpv0a1; Stv1; Vph1; Vpp-1; VPP1
UniProt ID: (Human) Q93050, (Mouse) Q9Z1G4, (Rat) P25286
Entrez Gene ID: (Human) 535, (Mouse) 11975, (Rat) 29757
Molecular Function:
ATP synthase
cation transporter
hydrolase
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.