Invitrogen
Manufacturer Code:PA518205
Catalog # PIPA518205
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (SIIYRMTNQKIRMD) corresponding to the C-terminus amino acids of ATP6AP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: (pro)renin receptor; ATP6AP2 ATP6IP2 ATPase H+ transporting lysosomal interacting protein 2 M8-9 APT6M8-9 ATP6M8-9 renin receptor V-ATPase M8.9 subunit ATPase membrane sector associated protein M8-9 vacuolar ATP synthase membrane sector associated protein M8-9 ATPase H+ transporting lysosomal (vacuolar proton pump) membrane sector associated protein M8-9 ELDF10 HT028 MGC99577 MRXE MSTP009 XMRE ATPase H+ transporting lysosomal interacting protein 2 embryonic liver differentiation factor 10; ATP6M8-9; ATPase H(+)-transporting lysosomal accessory protein 2; ATPase H(+)-transporting lysosomal-interacting protein 2; ATPase, H(+)-transporting, lysosomal-interacting protein 2; ATPase, H+ transporting, lysosomal (vacuolar proton pump) membrane sector associated protein M8-9; ATPase, H+ transporting, lysosomal accessory protein 2; ATPase, H+ transporting, lysosomal interacting protein 1; ATPase, H+ transporting, lysosomal interacting protein 2; Embryonic liver differentiation factor 10; ER-localized type I transmembrane adapter; ER-localized type I transmembrane adaptor; N14F; prorenin receptor; Renin receptor; Renin receptor C-terminal fragment; Renin receptor cytoplasmic fragment; Renin receptor extracellular fragment; Renin receptor N-terminal fragment; Renin/prorenin receptor; V-ATPase M8.9 subunit; Vacuolar ATP synthase membrane sector-associated protein M8-9; vacuolar proton ATP synthase membrane sector associated protein M8-9
Gene Aliases: (P)RR; 5730403E06Rik; APT6M8-9; ATP6AP2; ATP6IP2; ATP6M8-9; CAPER; ELDF10; HT028; M8-9; MRXE; MRXSH; MSTP009; PRR; PSEC0072; RENR; XMRE; XPDS
UniProt ID: (Human) O75787, (Rat) Q6AXS4, (Mouse) Q9CYN9
Entrez Gene ID: (Human) 10159, (Rat) 302526, (Mouse) 70495
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.