Abnova
Manufacturer Code:H00000537M01
Catalog # 89-016-284
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A2 |
Immunogen | ATP6AP1 (NP_001174 51 a.a. ~ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant ATP6AP1.
For Research Use Only
Protein Aliases: 16A; Ac45; ATP6IP1; ATP6S1; ATPase, H+ transporting, lysosomal (vacuolar proton pump), subunit 1; ATPase, H+ transporting, lysosomal accessory protein 1; ATPase, H+ transporting, lysosomal interacting protein 1; CF2; H-ATPase subunit; MGC129781; Protein XAP-3; V-ATPase Ac45 subunit; V-ATPase S1 accessory protein; V-ATPase subunit S1; V-type proton ATPase subunit S1; Vacuolar proton pump subunit S1; VATPS1; XAP-3; XAP3
Gene Aliases: 16A; Ac45; ATP6AP1; ATP6IP1; ATP6S1; CF2; VATPS1; XAP-3; XAP3
UniProt ID: (Human) Q15904
Entrez Gene ID: (Human) 537
Molecular Function:
ATP synthase
cation transporter
hydrolase
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.