Novus Biologicals
Manufacturer Code:NBP157020
Catalog # NBP157020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to ATP6AP1 (ATPase H+ transporting lysosomal accessory protein 1) The peptide sequence was selected from the middle region of ATP6AP1. Peptide sequence SPVIHPPVSYNDTAPRILFWAQNFSVAYKDQWEDLTPLTFGVQELNLTGS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 16A Ac45 ATP6IP1V-ATPase subunit S1 ATP6S1ORF ATPase H+ transporting lysosomal (vacuolar proton pump) subunit 1 ATPase H+ transporting lysosomal accessory protein 1 H+ transporting lysosomal interacting protein 1 H-ATPase subunit MGC129781 Protein XAP-3 Vacuolar proton pump subunit S1 VATPS1V-ATPase Ac45 subunit V-type proton ATPase subunit S1 XAP-3 XAP3V-ATPase S1 accessory protein; ATPase, H+ transporting, lysosomal (vacuolar proton pump), subunit 1; ATPase, H+ transporting, lysosomal accessory protein 1; ATPase, H+ transporting, lysosomal interacting protein 1; H-ATPase subunit; Protein XAP-3; V-ATPase Ac45 subunit; V-ATPase S1 accessory protein; V-ATPase subunit S1; V-type proton ATPase subunit S1; Vacuolar proton pump subunit S1
Gene Aliases: 16A; Ac45; ATP6AP1; ATP6IP1; ATP6S1; CF2; VATPS1; XAP-3; XAP3
UniProt ID: (Human) Q15904
Entrez Gene ID: (Human) 537
Molecular Function: ATP synthase cation transporter hydrolase transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.