Abnova
Manufacturer Code:H00000522M02
Catalog # 89-016-277
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2F4 |
Immunogen | ATP5J (AAH01178 1 a.a. ~ 108 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant ATP5J.
For Research Use Only
Protein Aliases: ATP synthase peripheral stalk subunit F6; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F6; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit F6; ATP synthase-coupling factor 6, mitochondrial; ATP5; ATP5A; ATPase subunit F6; ATPM; CF6; coupling factor 6; F6; mitochondrial ATP synthase, coupling factor 6; mitochondrial ATP synthase, subunit F6; mitochondrial ATPase coupling factor 6; proliferation-inducing protein 36
Gene Aliases: ATP5; ATP5A; ATP5J; ATP5PF; ATPM; CF6; F6
UniProt ID: (Human) P18859
Entrez Gene ID: (Human) 522
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.