Invitrogen
Manufacturer Code:PA556630
Catalog # PIPA556630
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ATP5J |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP synthase peripheral stalk subunit F6; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit F6; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit F6; ATP synthase-coupling factor 6, mitochondrial; ATP5 ATP5A ATPM CF6 F6 ATP synthase H+ transporting mitochondrial F0 complex subunit F6 ATP synthase-coupling factor 6 mitochondrial ATPase subunit F6 coupling factor 6 mitochondrial ATP synthase coupling factor 6 mitochondrial ATP synthase subunit F6 mitochondrial ATPase; ATPase subunit F6; coupling factor 6; mitochondrial ATP synthase, coupling factor 6; mitochondrial ATP synthase, subunit F6; mitochondrial ATPase coupling factor 6; proliferation-inducing protein 36
Gene Aliases: ATP5; ATP5A; ATP5J; ATP5PF; ATPM; CF6; F6
UniProt ID: (Human) P18859
Entrez Gene ID: (Human) 522
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.