Proteintech
Manufacturer Code:164831AP150UL
Catalog # 50-173-0977
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ATP5I Fusion Protein Ag9605 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP synthase e chain, mitochondrial; ATP synthase membrane subunit e; ATP synthase subunit e, mitochondrial; ATP synthase subunit e, mitochondrial, N-terminally processed; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit E; ATP synthase, H+ transporting, mitochondrial F1F0 complex, subunit e; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit E; ATP5I ATP5K ATPase subunit e; ATPase subunit e; F1F0-ATP synthase, murine e subunit
Gene Aliases: 2610008D24Rik; ATP5I; ATP5K; ATP5ME; Lfm-1; Lfm1
UniProt ID: (Human) P56385, (Rat) P29419, (Mouse) Q06185
Entrez Gene ID: (Human) 521, (Rat) 140608, (Mouse) 11958
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.