Proteintech
Manufacturer Code:175891AP150UL
Catalog # 50-173-1668
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ATP5H Fusion Protein Ag11429 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP synthase D chain, mitochondrial; ATP synthase peripheral stalk subunit d; ATP synthase subunit d, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit d; ATP synthase, H+ transporting, mitochondrial F1F0, subunit d; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit d; ATP5H ATP5JD ATPase subunit d ATPQ; ATPase subunit d; My032 protein
Gene Aliases: 0610009D10Rik; ATP5H; Atp5jd; ATP5PD; ATPQ; My032
UniProt ID: (Human) O75947, (Mouse) Q9DCX2, (Rat) P31399
Entrez Gene ID: (Human) 10476, (Mouse) 71679, (Rat) 641434
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.