Invitrogen
Manufacturer Code:PA597484
Catalog # PIPA597484
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human ATP synthase subunit delta mitochondrial protein (1-168AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP synthase F1 subunit delta; ATP synthase subunit delta, mitochondrial; F-ATPase delta subunit; mitochondrial ATP synthase complex delta-subunit precusor; mitochondrial ATP synthase, delta subunit
Gene Aliases: ATP5D; ATP5F1D
UniProt ID: (Human) P30049
Entrez Gene ID: (Human) 513
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.