Novus Biologicals
Manufacturer Code:NBP300344FR
Catalog # NB087355
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant ATP5D (P30049 Ala23-Glu168). |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP synthase F1 subunit delta; ATP synthase subunit delta mitochondrial ATP synthase H+ transporting mitochondrial F1 complex delta subunit F-ATPase delta subunit mitochondrial ATP synthase complex delta-subunit precusor mitochondrial ATP synthase delta subunit; ATP synthase subunit delta, mitochondrial; F-ATPase delta subunit; mitochondrial ATP synthase complex delta-subunit precusor; mitochondrial ATP synthase, delta subunit
Gene Aliases: ATP5D; ATP5F1D
UniProt ID: (Human) P30049
Entrez Gene ID: (Human) 513
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.