Novus Biologicals
Manufacturer Code:NBP215512
Catalog # NBP215512
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human ATP5A1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP synthase alpha chain, mitochondrial; ATP synthase F1 subunit alpha; ATP synthase H+ transporting mitochondrial F1 complex alpha subunit 1 ATP synthase H+ transporting mitochondrial F1 complex alpha subunit isoform1 cardiac muscle ATP synthase H+ transporting mitochondrial F1 complex alpha subunit isoform2 non-cardiac muscle-like 2 ATP sythase (F1-ATPase) alpha subunit ATP5AL2 ATP5AMOM2 ATPMATP synthase subunit alpha mitochondrial cardiac muscle EC 3.6.3 EC 3.6.3.14 hATP1 mitochondrial ATP synthetase oligomycin-resistant OMR ORMATP synthase alpha chain mitochondrial; ATP synthase subunit alpha, mitochondrial; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform 1, cardiac muscle; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform 2, non-cardiac muscle-like 2; ATP sythase (F1-ATPase) alpha subunit; epididymis secretory sperm binding protein Li 123m; mitochondrial ATP synthetase, oligomycin-resistant
Gene Aliases: ATP5A; ATP5A1; ATP5AL2; ATP5F1A; ATPM; COXPD22; hATP1; HEL-S-123m; MC5DN4; MOM2; OMR; ORM
UniProt ID: (Human) P25705
Entrez Gene ID: (Human) 498
Molecular Function: ATP synthase DNA binding protein anion channel cation transporter hydrolase ion channel ligand-gated ion channel nucleic acid binding receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.