Invitrogen
Manufacturer Code:PA525105
Catalog # PIPA525105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 52-78 amino acids from the N-terminal region of human ATP4B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP6B ATPase H+/K+ transporting beta polypeptide gastric H(+)/K(+) ATPase subunit beta gastric H+/K+ ATPase beta subunit gastric hydrogen-potassium ATPase beta potassium-transporting ATPase beta chain potassium-transporting ATPase subunit beta proton pump be; ATPase, H+/K+ exchanging, beta polypeptide; ATPase, H+/K+ transporting, beta polypeptide; ATPase, H+/K+ transporting, beta polypeptide, gastric specific; Gastric H(+)/K(+) ATPase subunit beta; gastric H+/K+ ATPase beta subunit; gastric hydrogen-potassium ATPase, beta; H+,K+-ATPase; H+/K+-ATPase beta; H,K-ATPase-Beta; potassium-transporting ATPase beta chain; Potassium-transporting ATPase subunit beta; Proton pump beta chain
Gene Aliases: ATP4B; ATP6B; AV080843
UniProt ID: (Human) P51164, (Mouse) P50992
Entrez Gene ID: (Human) 496, (Mouse) 11945
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.