Proteintech
Manufacturer Code:186921AP150UL
Catalog # 50-173-2190
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ATP4B Fusion Protein Ag6935 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP4B ATP6B H+/K+ ATPase beta Proton pump beta chain; ATPase H+K+-transporting beta / (gastric HK-ATPase beta subunit) defined by SSR; ATPase, H+,K+-transporting, beta / (gastric H,K-ATPase beta subunit), defined by SSR; ATPase, H+/K+ exchanging, beta polypeptide; ATPase, H+/K+ transporting, beta polypeptide; ATPase, H+/K+ transporting, beta polypeptide, gastric specific; Gastric H(+)/K(+) ATPase subunit beta; gastric H+/K+ ATPase beta subunit; gastric hydrogen-potassium ATPase, beta; H+,K+-ATPase; H+/K+-ATPase beta; H,K-ATPase-Beta; potassium-transporting ATPase beta chain; Potassium-transporting ATPase subunit beta; Proton pump beta chain
Gene Aliases: ATP4B; ATP6B; AV080843; S76401S1
UniProt ID: (Human) P51164, (Rat) P18598, (Mouse) P50992
Entrez Gene ID: (Human) 496, (Rat) 24217, (Mouse) 11945
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.