Abnova
Manufacturer Code:H00000493M03A
Catalog # 89-104-127
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C7 |
Immunogen | ATP2B4 (NP_0016751 a.a. &bsim 92 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant ATP2B4.
For Research Use Only
Protein Aliases: ATP2B2; ATPase, Ca++ transporting, plasma membrane 4; DKFZp686G08106; DKFZp686M088; Matrix-remodeling-associated protein 1; MXRA1; Plasma membrane calcium ATPase isoform 4; Plasma membrane calcium pump isoform 4; Plasma membrane calcium-transporting ATPase 4; PMCA4; PMCA4b; PMCA4x; sarcolemmal calcium pump
Gene Aliases: ATP2B2; ATP2B4; MXRA1; PMCA4; PMCA4b; PMCA4x
UniProt ID: (Human) P23634
Entrez Gene ID: (Human) 493
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.