Abnova
Manufacturer Code:H00000490M01
Catalog # 89-016-272
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3E2 |
Immunogen | ATP2B1 (NP_001673 1 a.a. ~ 97 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant ATP2B1.
For Research Use Only
Protein Aliases: ATPase, Ca++ transporting, plasma membrane 1; Plasma membrane calcium ATPase isoform 1; plasma membrane calcium pump; Plasma membrane calcium pump isoform 1; Plasma membrane calcium-transporting ATPase 1; PMCA1; PMCA1kb
Gene Aliases: ATP2B1; PMCA1; PMCA1kb
UniProt ID: (Human) P20020
Entrez Gene ID: (Human) 490
Molecular Function:
cation transporter
hydrolase
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.