Invitrogen
Manufacturer Code:MA529047
Catalog # PIMA529047
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 108 |
Immunogen | Recombit Human ATP1B4 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: ATP1B4; ATPase, Na+/K+ transporting, beta 4 polypeptide; BetaM; Na,K-ATPase beta m-subunit; Protein ATP1B4; X,K-ATPase beta-m subunit; X,K-ATPase subunit beta-m; X/potassium-transporting ATPase subunit beta-m
Gene Aliases: ATP1B4
UniProt ID: (Human) Q9UN42
Entrez Gene ID: (Human) 23439
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.