Invitrogen
Manufacturer Code:PA569429
Catalog # PIPA569429
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human ATP1B4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP1B4; ATPase, (Na+)/K+ transporting, beta 4 polypeptide; ATPase, Na+/K+ transporting, beta 4 polypeptide; BetaM; Na,K-ATPase beta m-subunit; Protein ATP1B4; X,K-ATPase beta-m subunit; X,K-ATPase subunit beta-m; X/potassium-transporting ATPase subunit beta-m
Gene Aliases: 1110020B02Rik; Ac2-628; ATP1B4
UniProt ID: (Bovine) A7MB71, (Human) Q9UN42, (Mouse) Q99ME6, (Rat) Q9R193
Entrez Gene ID: (Horse) 100055231, (Rabbit) 100350113, (Bovine) 510026, (Human) 23439, (Dog) 481036, (Mouse) 67821, (Rat) 84396, (Guinea pig) 100713904
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.