Abnova
Manufacturer Code:H00000483M05
Catalog # 89-016-261
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2F4 |
Immunogen | ATP1B3 (AAH11835 1 a.a. ~ 279 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant ATP1B3.
For Research Use Only
Protein Aliases: ATPB-3; CD298; FLJ29027; Sodium/potassium-dependent ATPase subunit beta-3; Sodium/potassium-transporting ATPase subunit beta-3
Gene Aliases: ATP1B3; ATPB-3; CD298
UniProt ID: (Human) P54709
Entrez Gene ID: (Human) 483
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.