Proteintech
Manufacturer Code:111421AP150UL
Catalog # 50-172-6912
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ATP1B3 Fusion Protein Ag1589 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP1B3 ATPB 3 CD298; ATPase, Na+/K+ beta 3 polypeptide; ATPB-3; CD298; Na K-ATPase beta-3 subunit; Sodium/potassium-dependent ATPase subunit beta-3; Sodium/potassium-transporting ATPase subunit beta-3
Gene Aliases: AA409958; AI664000; ATP1B3; ATPB-3; AW212096; CD298
UniProt ID: (Human) P54709, (Mouse) P97370
Entrez Gene ID: (Human) 483, (Mouse) 11933
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.