Proteintech
Manufacturer Code:223381AP150UL
Catalog # 50-173-3836
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ATP1B2 Fusion Protein Ag17818 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adhesion molecule in glia; adhesion molecule on glia; AMOG; AMOG ATP1B2 ATPase Na+/K+ beta 2; ATPase, Na+/K+ transporting, beta 2 polypeptide; ATPase, Na+K+ transporting, beta polypeptide 2; glial cell adhesion molecule; Na, K-ATPase beta-2 polypeptide; snoRNA MBI-85; sodium pump subunit beta-2; sodium-potassium ATPase subunit beta 2 (non-catalytic); sodium/potassium-dependent ATPase beta-2 subunit; Sodium/potassium-dependent ATPase subunit beta-2; sodium/potassium-transporting ATPase beta-2 chain; Sodium/potassium-transporting ATPase subunit beta-2
Gene Aliases: AMOG; ATP1B2; Atpb-2; ATPB2; ATPB2S; RATATPB2S
UniProt ID: (Human) P14415, (Mouse) P14231
Entrez Gene ID: (Human) 482, (Rat) 24214, (Mouse) 11932
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.