Invitrogen
Manufacturer Code:PA529719
Catalog # PIPA529719
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 83 and 303 of Human Sodium Potassium ATPase Beta 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenosinetriphosphatase; ATP1B Beta 1-subunit of Na(+)K(+)-ATPase Na K-ATPase beta-1 polypeptide adenosinetriphosphatase sodium/potassium-dependent ATPase beta-1 subunit sodium/potassium-transporting ATPase beta-1 chain sodium/potassium-transporting ATPase subunit beta-1; ATPase, Na+/K+ transporting, beta 1 polypeptide; Beta 1-subunit of Na(+),K(+)-ATPase; Na, K-ATPase beta-1 polypeptide; Na,K-ATPase beta 1 subunit; sodium pump subunit beta-1; sodium-potassium ATPase subunit beta 1 (non-catalytic); sodium/potassium ATPase beta subunit; sodium/potassium-dependent ATPase beta-1 subunit; Sodium/potassium-dependent ATPase subunit beta-1; sodium/potassium-transporting ATPase beta-1 chain; Sodium/potassium-transporting ATPase subunit beta-1
Gene Aliases: ATP1B; ATP1B1; Atp4b; Atpb; Atpb-1; ATPBS; NKbeta1
UniProt ID: (Human) P05026, (Rat) P07340, (Mouse) P14094
Entrez Gene ID: (Human) 481, (Rat) 25650, (Mouse) 11931
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.