Invitrogen
Manufacturer Code:MA537905
Catalog # PIMA537905
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2458 |
Immunogen | Recombinant protein of human ATP1A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Atp1a2 Atpa-3 ATPase Na+/K+ transporting alpha 2 polypeptide ATPase Na+/K+ transporting subunit alpha 2 ATPase Na+/K+ transporting alpha 2 polypeptide ATPase Na+K+ transporting alpha 2 AW060654 FHM2 KIAA0778 MHP2 mKIAA0778 Na(+)/K(+) ATPase alpha(+) subunit Na(+)/K(+) ATPase alpha-2 subunit Na+/K+ -ATPase alpha 2 subunit Na+/K+ ATPase alpha-A(+) catalytic polypeptide Na+/K+ ATPase alpha-B polypeptide RATATPA2 Sodium Potassium ATPase Sodium pump subunit alpha-2 sodium/potassium-transporting ATPase alpha-2 chain sodium/potassium-transporting ATPase subunit alpha-2 sodium-potassium ATPase catalytic subunit alpha-2; ATPase Na+/K+ transporting alpha 2 polypeptide; ATPase, Na+/K+ transporting, alpha 2 polypeptide; ATPase, Na+K+ transporting, alpha 2; Na(+)/K(+) ATPase alpha(+) subunit; Na(+)/K(+) ATPase alpha-2 subunit; Na+/K+ -ATPase alpha 2 subunit; Na+/K+ ATPase, alpha-A(+) catalytic polypeptide; Na+/K+ ATPase, alpha-B polypeptide; Sodium pump subunit alpha-2; sodium-potassium ATPase catalytic subunit alpha-2; sodium/potassium-transporting ATPase alpha-2 chain; Sodium/potassium-transporting ATPase subunit alpha-2
Gene Aliases: ATP1A2; Atpa-3; AW060654; FHM2; KIAA0778; MHP2; mKIAA0778; RATATPA2
UniProt ID: (Human) P50993, (Rat) P06686, (Mouse) Q6PIE5
Entrez Gene ID: (Human) 477, (Rat) 24212, (Mouse) 98660
Molecular Function: cation transporter hydrolase ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.